Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163622.1 | NP_001157094.1 | 6 | 1204 | ||
NM_001163623.1 | NP_001157095.1 | 5 | 668 | |||
NM_001163624.1 | NP_001157096.1 | 5 | 536 | |||
NM_145984.3 | NP_666096.3 | 6 | 768 | |||
GenBank mRNA | AK028877.1 | - | 6 | 1162 | ||
AK050878.1 | - | 5 | 530 | |||
AK083070.1 | - | 6 | 709 | |||
AK163108.1 | - | 6 | 1169 | |||
BC004612.1 | - | 5 | 622 | 3585402 |