Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005765.2 | NP_005756.2 | 7 | 725 | ||
GenBank mRNA | AF109363.1 | - | 7 | 687 | ||
AF248966.1 | - | 7 | 730 | |||
AF291814.1 | - | 7 | 712 | |||
AK075382.1 | - | 7 | 783 | |||
AK293510.1 | - | 5 | 473 | |||
AK296614.1 | - | 6 | 588 | |||
AK297007.1 | - | 8 | 584 | |||
AK297760.1 | - | 6 | 699 | |||
AK314633.1 | - | 8 | 759 | |||
AK315948.1 | - | 6 | 560 | |||
AL049929.1 | - | 6 | 565 | |||
AY038990.1 | - | 7 | 728 | |||
AY429341.1 | - | 6 | 588 | |||
BC010395.1 | - | 3 | 185 | |||
BC084541.1 | - | 7 | 705 | 6575732 | ||
Y17975.1 | - | 2 | 64 |