Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146017.3 | NP_666129.1 | 7, 8 | 878 | ||
GenBank mRNA | AK028946.1 | - | 7, 8 | 878 | ||
AK160577.1 | - | 7, 8 | 882 | |||
BC023693.1 | - | 7, 8 | 865 | 5344479 | ||
BC025550.1 | - | 7, 8 | 823 | 5320737 | ||
BC027245.1 | - | 7, 8 | 841 | 3602400 | ||
BC027839.1 | - | 3, 4 | 430 | |||
BC031196.1 | - | 7, 8 | 852 | 5059564 |