Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146020.1 | NP_666132.1 | 3 | 1028 | ||
GenBank mRNA | AK050107.1 | - | 3 | 1042 | ||
AK050212.1 | - | 3 | 1042 | |||
BC025515.1 | - | 3 | 1027 | 5309987 | ||
BC030735.1 | - | 3 | 1028 | 5126172 |