Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctage5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165253.1 | NP_001158725.1 | 5 | 484 | ||
NM_001165254.1 | NP_001158726.1 | 5 | 483 | |||
NM_146034.3 | NP_666146.3 | 7 | 582 | |||
GenBank mRNA | AK029466.1 | - | 5 | 483 | ||
AK053010.1 | - | 6 | 489 | |||
AK080312.1 | - | 6 | 531 | |||
AK087188.1 | - | 6 | 497 | |||
AK157358.1 | - | 5 | 485 | |||
BC026864.1 | - | 7 | 534 | 2647214 | ||
BQ571563.1 | - | 5 | 483 |