Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146054.2 | NP_666166.2 | 15 | 2082 | ||
GenBank mRNA | AK145030.1 | - | 15 | 2024 | ||
AK165608.1 | - | 15 | 2052 | |||
AK166552.1 | - | 15 | 2021 | |||
BC033436.1 | - | 15 | 2003 | 4950810 | ||
BC034168.1 | - | 6 | 736 | |||
CN716799.1 | - | 4 | 481 |