Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146067.3 | NP_666179.2 | 2 | 187 | ||
GenBank mRNA | AK029379.1 | - | 2 | 111 | ||
AK029442.1 | - | 2 | 210 | |||
AK032288.1 | - | 2 | 150 | |||
AK041329.1 | - | 2 | 160 | |||
AK049721.1 | - | 2 | 188 | |||
AK133977.1 | - | 2 | 180 | |||
AK134718.1 | - | 2 | 188 | |||
BC025476.1 | - | 2 | 108 | |||
BC034347.1 | - | 2 | 166 | 2654874 |