Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfb1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146074.1 | NP_666186.1 | 3 | 375 | ||
GenBank mRNA | AF508971.1 | - | 3 | 375 | ||
BC032930.1 | - | 3 | 375 | 1382837 |