Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195485.1 | NP_001182414.1 | 8 | 872 | ||
NM_001195486.1 | NP_001182415.1 | 8 | 848 | |||
NM_001195487.1 | NP_001182416.1 | 7 | 836 | |||
NM_146083.2 | NP_666195.1 | 8 | 881 | |||
GenBank mRNA | AK034952.1 | - | 9 | 1334 | ||
AK045884.1 | - | 8 | 1043 | |||
AK082848.1 | - | 8 | 872 | |||
AK137795.1 | - | 8 | 882 | |||
AK161652.1 | - | 8 | 880 | |||
AK168355.1 | - | 8 | 881 | |||
AK178246.1 | - | 3 | 202 | |||
BC025529.1 | - | 8 | 872 | 5342587 | ||
BC027391.1 | - | 7 | 1734 | 4945988 |