Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146097.3 | NP_666209.1 | 4 | 468 | ||
GenBank mRNA | AK076027.1 | - | 4 | 423 | ||
AK132291.1 | - | 4 | 454 | |||
BC018472.1 | - | 4 | 410 | 3661842 | ||
BC053745.1 | - | 4 | 503 | 6505520 |