Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG3 SIRNA Assays ›
Gene Symbol:
ALG3
Gene Name:
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases:
CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
Chromosome Location:
Chr.3: 184242329 - 184249525 on Build GRCh38
UniGene ID:
Hs.478481
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17023
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006941.2
NP_001006942.1
7
1074
NM_005787.5
NP_005778.1
7
1001
NR_024533.1
6
901
NR_024534.1
7
964
XM_011512322.1
XP_011510624.1
7
886
XM_011512323.2
XP_011510625.1
7
872
GenBank mRNA
AK289361.1
-
7
1030
AK299575.1
-
6
892
BC002839.2
-
7
976
3636604
BC004313.1
-
7
956
BC021062.1
-
7
965
KP189279.1
-
7
994
KP189280.1
-
7
957
KP189281.1
-
6
810
KP189282.1
-
7
1111
KP189283.1
-
6
894
KP189284.1
-
7
992
KP189285.1
-
7
955
KP189286.1
-
6
810
KP189287.1
-
7
910
KP189288.1
-
6
892
KP189289.1
-
7
991
KP189290.1
-
7
955
KP189291.1
-
6
810
KP189292.1
-
7
910
KP189293.1
-
6
892
KP189294.1
-
7
970
KP189295.1
-
6
764
KP189296.1
-
7
933
KP189297.1
-
6
727
KP189298.1
-
4
523
KP189299.1
-
7
826
KP189300.1
-
6
870
KP189302.1
-
7
991
KP189303.1
-
7
953
KP189304.1
-
6
810
KP189305.1
-
7
910
KP189306.1
-
6
890
KP189307.1
-
7
990
KP189308.1
-
7
953
KP189309.1
-
6
810
KP189310.1
-
7
910
KP189311.1
-
6
890
KP189312.1
-
7
990
KP189313.1
-
6
784
KP189315.1
-
7
953
KP189316.1
-
6
747
KP189318.1
-
6
810
KP189319.1
-
5
604
KP189321.1
-
7
910
KP189322.1
-
6
704
KP189324.1
-
6
890
KP189325.1
-
5
684
Y09022.1
-
7
1001
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
dolichol-linked oligosaccharide biosynthetic process
mannosylation
alpha-1,3-mannosyltransferase activity
dol-P-Man:Man(5)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-nglqg:80/10.239.166.193:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0