Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153057.4 | NP_694697.3 | 7 | 725 | ||
GenBank mRNA | AK030584.1 | - | 7 | 692 | ||
AK035957.1 | - | 7 | 695 | |||
AK080754.1 | - | 7 | 584 | |||
AK082495.1 | - | 7 | 604 | |||
AK082963.1 | - | 7 | 584 | |||
AK142338.1 | - | 7 | 694 | |||
AK150107.1 | - | 7 | 679 | |||
AK165643.1 | - | 7 | 586 | |||
AK193074.1 | - | 2 | 110 | |||
AK193375.1 | - | 2 | 110 | |||
AK199535.1 | - | 2 | 110 | |||
BC022138.1 | - | 6 | 441 | |||
BC072630.1 | - | 7 | 671 | 6833516 |