Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153057.4 | NP_694697.3 | 9, 10 | 1083 | ||
GenBank mRNA | AK030584.1 | - | 9, 10 | 1050 | ||
AK035957.1 | - | 9, 10 | 1053 | |||
AK080754.1 | - | 9, 10 | 946 | |||
AK082495.1 | - | 9, 10 | 962 | |||
AK082963.1 | - | 9, 10 | 942 | |||
AK142338.1 | - | 9, 10 | 1053 | |||
AK150107.1 | - | 9, 10 | 1037 | |||
AK165643.1 | - | 9, 10 | 944 | |||
BC022138.1 | - | 8, 9 | 799 | |||
BC072630.1 | - | 9, 10 | 1029 | 6833516 |