Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081277.1 | NP_001074746.1 | 13 | 1764 | ||
GenBank mRNA | AB060081.1 | - | 4 | 376 | ||
AK053807.1 | - | 13 | 1762 |