Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153075.3 | NP_694715.2 | 12 | 2076 | ||
GenBank mRNA | AF411816.1 | - | 12 | 1819 | ||
AK142380.1 | - | 6 | 1004 | |||
BC034181.1 | - | 6 | 1136 | |||
BC132140.1 | - | 12 | 1723 | 40130417 |