Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251973.1 | NP_001238902.1 | 5 | 2844 | ||
NM_001251974.1 | NP_001238903.1 | 5 | 2797 | |||
NM_005822.3 | NP_005813.2 | 4 | 2919 | |||
XM_011514226.1 | XP_011512528.1 | 5 | 2886 | |||
GenBank mRNA | AK090990.1 | - | 5 | 2768 | ||
AY034085.1 | - | 5 | 2749 | |||
AY034086.1 | - | 5 | 2762 | |||
BP264912.1 | - | 1 | 558 | |||
D83407.1 | - | 4 | 2693 |