Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028000.1 | - | 1 | 31 | ||
AK052762.1 | - | 4 | 1334 | |||
AK085717.1 | - | 5 | 1496 | |||
AK154613.1 | - | 4 | 1480 | |||
AK163171.1 | - | 4 | 1427 | |||
AK170641.1 | - | 4 | 1466 | |||
AK180272.1 | - | 1 | 106 | |||
BC005765.1 | - | 3 | 1202 | |||
BC007169.1 | - | 1 | 359 | |||
BC022976.1 | - | 4 | 1412 | 5362925 | ||
BC024940.1 | - | 4 | 1409 | |||
BC071258.1 | - | 4 | 1345 |