Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E430025E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153548.2 | NP_705776.2 | 19 | 2556 | ||
GenBank mRNA | AK088754.1 | - | 4 | 429 | ||
AK128970.1 | - | 19 | 2571 | |||
AK129086.1 | - | 285 | ||||
AK163968.1 | - | 2 | 180 | |||
BC031364.1 | - | 5 | 570 | |||
BC034070.1 | - | 9 | 1156 | 4217923 | ||
BC040815.1 | - | 9 | 1156 | 4217923 | ||
BC067035.1 | - | 19 | 2556 | 30532658 |