Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172050.2 | NP_742047.1 | 4 | 923 | ||
GenBank mRNA | BC034097.1 | - | 4 | 948 | 4487018 | |
BC039971.1 | - | 4 | 959 | 4487018 |