Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172274.2 | NP_758478.1 | 9 | 1012 | ||
GenBank mRNA | AK077658.1 | - | 6 | 487 | ||
AK145128.1 | - | 6 | 550 | |||
BC032290.1 | - | 9 | 956 | 5369681 |