Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172523.3 | NP_766111.1 | 3 | 317 | ||
GenBank mRNA | AJ555564.1 | - | 2 | 181 | ||
AK033871.1 | - | 3 | 273 | |||
AK034368.1 | - | 2 | 538 | |||
AK035644.1 | - | 3 | 300 | |||
AK041274.1 | - | 3 | 269 | |||
BC078449.1 | - | 3 | 232 | 5705966 |