Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 8 | 1218 | ||
GenBank mRNA | AK030952.1 | - | 8 | 1207 | ||
AK033634.1 | - | 7 | 2041 | |||
AK037337.1 | - | 8 | 1237 | |||
AK048454.1 | - | 8 | 1195 | |||
AK138809.1 | - | 8 | 1219 | |||
AK154133.1 | - | 8 | 1246 | |||
AK154348.1 | - | 8 | 1246 | |||
AK164885.1 | - | 8 | 1188 | |||
AK216242.1 | - | 3 | 114 | |||
BC057943.1 | - | 8 | 1242 | |||
BC076606.1 | - | 8 | 1210 | 30625515 | ||
BC089191.1 | - | 8 | 1192 | 30473852 |