Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172648.3 | NP_766236.2 | 7 | 1463 | ||
GenBank mRNA | BC040425.1 | - | 7 | 1463 | 3466347 | |
M74124.1 | - | 6 | 1244 |
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 7 | 1627 | ||
NM_001301745.1 | NP_001288674.1 | 7 | 1621 | |||
GenBank mRNA | AK152278.1 | - | 7 | 1495 | ||
AK152683.1 | - | 7 | 1495 | |||
AK152786.1 | - | 7 | 1504 | |||
AK166217.1 | - | 7 | 1627 | |||
AV250999.1 | - | 1 | 74 | |||
M74123.1 | - | 7 | 1482 | |||
S62227.1 | - | 7 | 1482 |
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170853.1 | NP_001164324.1 | 1758 | |||
GenBank mRNA | AK087931.1 | - | 8 | 1583 | ||
AK136391.1 | - | 1760 | ||||
FJ594740.1 | - | 8 | 1657 |