Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173383.2 | NP_775559.2 | 4 | 1092 | ||
GenBank mRNA | AK141343.1 | - | 3 | 1685 | ||
AY321066.1 | - | 4 | 1044 | |||
BC034897.1 | - | 3 | 1427 | 3470697 |