Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256534.1 | NP_001243463.1 | 2, 3 | 481 | ||
NM_001287387.1 | NP_001274316.1 | 3, 4 | 335 | |||
NM_005984.4 | NP_005975.1 | 3, 4 | 500 | |||
GenBank mRNA | AK292313.1 | - | 3, 4 | 404 | ||
AK298204.1 | - | 3, 4 | 333 | |||
BC004980.1 | - | 3, 4 | 361 | 2957917 | ||
BC008061.2 | - | 3, 4 | 363 | 2958945 | ||
BC018590.1 | - | 3, 4 | 258 | |||
BG473148.1 | - | 2, 3 | 455 | |||
BP265446.1 | - | 3, 4 | 335 | |||
DR006840.1 | - | 3, 4 | 402 | |||
HM037273.1 | - | 2, 3 | 336 | |||
L75823.1 | - | 3, 4 | 409 | |||
L77567.1 | - | 3, 4 | 382 | |||
U25147.1 | - | 3, 4 | 384 |