Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289429.1 | NP_001276358.1 | 5 | 2171 | ||
NM_001289430.1 | NP_001276359.1 | 4 | 2070 | |||
NM_001289431.1 | NP_001276360.1 | 5 | 2244 | |||
NM_001289432.1 | NP_001276361.1 | 5 | 2118 | |||
NM_173735.3 | NP_776096.2 | 4 | 2017 | |||
GenBank mRNA | AK076398.1 | - | 1 | 1092 | ||
AK122546.1 | - | 4 | 1946 | |||
AK154476.1 | - | 4 | 1954 | |||
AK160481.1 | - | 5 | 2171 | |||
AK160643.1 | - | 4 | 1943 | |||
BC026384.1 | - | 4 | 1949 | 4168224 |