Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175445.4 | NP_780654.1 | 11 | 1463 | ||
GenBank mRNA | AK028384.1 | - | 11 | 1367 | ||
AK172906.1 | - | 11 | 1269 | |||
BC057402.1 | - | 11 | 1285 | 5698333 |