Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175466.4 | NP_780675.1 | 2 | 1402 | ||
GenBank mRNA | AK032578.1 | - | 2 | 1403 | ||
AK164226.1 | - | 2 | 1385 | |||
BC137653.1 | - | 1 | 1315 | 8860672 | ||
BC137654.1 | - | 1 | 1315 | 8860673 |