Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175467.3 | NP_780676.1 | 6 | 917 | ||
GenBank mRNA | AK048374.1 | - | 5 | 778 | ||
AK054240.1 | - | 5 | 1004 | |||
AK078679.1 | - | 5 | 981 | |||
AK078686.1 | - | 6 | 918 | |||
BC094496.1 | - | 5 | 763 | 3483569 |