Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172439.1 | NP_001165910.1 | 4 | 521 | ||
NM_001172440.1 | NP_001165911.1 | 2 | 332 | |||
NM_006025.3 | NP_006016.1 | 3 | 398 | |||
GenBank mRNA | AK075446.1 | - | 4 | 485 | ||
AK295963.1 | - | 1 | 120 | |||
AK300169.1 | - | 2 | 294 | |||
AK302308.1 | - | 1 | 112 | |||
AX772808.1 | - | 4 | 473 | |||
BC069715.1 | - | 3 | 277 | 7262649 | ||
BC074763.2 | - | 3 | 283 | 30915175 | ||
BC111782.1 | - | 3 | 372 | 40037008 | ||
DA838842.1 | - | 3 | 399 | |||
DA841959.1 | - | 3 | 398 | |||
DC381238.1 | - | 2 | 333 | |||
M32402.1 | - | 3 | 393 | |||
M36109.1 | - | 3 | 393 |