Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldoc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177630.3 | NP_808298.2 | 1 | 4103 | ||
GenBank mRNA | AK083881.1 | - | 1 | 4065 | ||
BC023053.1 | - | 1 | 530 | |||
BC035491.1 | - | 1 | 3159 | |||
BC058638.1 | - | 1 | 3772 | 6418256 | ||
BC094559.1 | - | 1 | 4103 | 6826763 |