Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027712.3 | NP_081988.3 | 2 | 938 | ||
NM_177639.6 | NP_808307.2 | 2 | 938 | |||
GenBank mRNA | AK031410.1 | - | 6 | 1628 | ||
AK033901.1 | - | 5 | 1471 | |||
AK035763.1 | - | 5 | 1489 | |||
AK043683.1 | - | 5 | 1526 | |||
AK147448.1 | - | 6 | 1755 | |||
AK220515.1 | - | 5 | 1818 | |||
AY243846.2 | - | 1 | 773 | |||
BC062120.1 | - | 2 | 939 | 6822047 | ||
BC094369.1 | - | 1 | 147 |