Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016752.1 | NP_058032.1 | 7, 8 | 860 | ||
GenBank mRNA | AK002961.1 | - | 7, 8 | 854 | ||
AK007711.1 | - | 7, 8 | 866 | |||
AK160124.1 | - | 7, 8 | 927 | |||
BC002029.1 | - | 7, 8 | 841 | 3592141 | ||
D87990.1 | - | 7, 8 | 860 |