Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035122.2 | NP_001030294.1 | 10 | 3075 | ||
NM_027307.4 | NP_081583.3 | 11 | 3211 | |||
GenBank mRNA | AK051066.1 | - | 1 | 1157 | ||
AK133865.1 | - | 10 | 3077 | |||
BC011152.1 | - | 10 | 3037 | 4188434 | ||
BC018170.1 | - | 1 | 77 | |||
BC098486.1 | - | 11 | 3207 | 30024953 |