Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033073.3 | NP_149064.1 | 4, 5 | 741 | ||
GenBank mRNA | AF509888.1 | - | 4, 5 | 718 | ||
AK002434.1 | - | 4, 5 | 719 | |||
AK137217.1 | - | 719 | ||||
AK146914.1 | - | 4, 5 | 719 | |||
BC010337.1 | - | 3, 4 | 2735 | |||
BC030403.1 | - | 4, 5 | 752 | 5355662 |