Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053261.2 | NP_444491.1 | 7 | 923 | ||
GenBank mRNA | AF353730.1 | - | 7 | 922 | ||
AK154489.1 | - | 7 | 961 | |||
AK154845.1 | - | 7 | 894 | |||
AK170962.1 | - | 7 | 961 | |||
AK171626.1 | - | 7 | 896 | |||
BC011093.1 | - | 7 | 898 | 4012163 |