Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054094.5 | NP_473435.1 | 3 | 291 | ||
GenBank mRNA | AB059428.1 | - | 3 | 295 | ||
AK149550.1 | - | 3 | 301 | |||
AK149586.1 | - | 3 | 317 | |||
BC016414.1 | - | 3 | 288 | 4238964 | ||
BC022149.1 | - | 3 | 269 | 5102652 |