Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130450.2 | NP_569717.1 | 5 | 1465 | ||
GenBank mRNA | AB072039.1 | - | 5 | 1414 | ||
AF480860.1 | - | 5 | 1384 | |||
AK029029.1 | - | 5 | 1467 | |||
AK087440.1 | - | 4 | 1337 | |||
AK165847.1 | - | 4 | 1490 | |||
AK167373.1 | - | 5 | 1431 | |||
AY053453.1 | - | 5 | 1446 | |||
BC098492.1 | - | 4 | 1460 | 30091954 | ||
BC100576.1 | - | 5 | 1403 | 30097278 |