Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285920.1 | NP_001272849.1 | 6 | 819 | ||
NM_001285921.1 | NP_001272850.1 | 6 | 802 | |||
NM_001285922.1 | NP_001272851.1 | 5 | 733 | |||
NM_001285923.1 | NP_001272852.1 | 5 | 733 | |||
NM_133201.3 | NP_573464.2 | 5 | 736 | |||
GenBank mRNA | AB048831.1 | - | 3 | 337 | ||
AF384100.1 | - | 6 | 616 | |||
AK042080.1 | - | 5 | 517 | |||
AK049583.1 | - | 5 | 526 | |||
AK052689.1 | - | 5 | 517 | |||
AK122220.1 | - | 5 | 597 | |||
AK146801.1 | - | 6 | 579 | |||
AK147863.1 | - | 5 | 532 | |||
AK169373.1 | - | 5 | 521 | |||
AY123975.1 | - | 5 | 683 | |||
BC046503.1 | - | 5 | 514 | 4191179 |