Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133205.3 | NP_573468.1 | 17 | 1257 | ||
GenBank mRNA | AF156979.1 | - | 17 | 1198 | ||
BC017145.1 | - | 16 | 1590 | 4506006 |