Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133207.2 | NP_573470.2 | 3 | 646 | ||
GenBank mRNA | AJ291608.1 | - | 3 | 453 | ||
AK034003.1 | - | 3 | 647 | |||
AK052366.1 | - | 3 | 530 |