Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133998.3 | NP_598759.1 | 6 | 2200 | ||
GenBank mRNA | AF391115.1 | - | 6 | 2172 | ||
AK088306.1 | - | 6 | 2168 | |||
BB007527.2 | - | 1 | 418 | |||
BC022775.1 | - | 3 | 1631 | |||
BC034216.1 | - | 6 | 2146 | 5008732 | ||
BI736563.1 | - | 1 | 367 |