Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930427A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134041.3 | NP_598802.2 | 8 | 1371 | ||
GenBank mRNA | AK029651.1 | - | 8 | 1373 | ||
AK167092.1 | - | 8 | 1330 | |||
AK206782.1 | - | 1 | 170 | |||
BC116830.1 | - | 4 | 710 | |||
BC116834.1 | - | 4 | 710 |