Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134054.2 | NP_598815.2 | 1, 2 | 196 | ||
GenBank mRNA | AK003171.1 | - | 1, 2 | 133 | ||
AK008904.1 | - | 1, 2 | 153 | |||
AK018380.1 | - | 1, 2 | 173 | |||
AK137543.1 | - | 1, 2 | 171 | |||
AK182833.1 | - | 1, 2 | 103 | |||
BC022674.1 | - | 1, 2 | 122 | 4236922 | ||
BY072251.1 | - | 1, 2 | 197 |