Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 10 | 1117 | ||
NM_134083.4 | NP_598844.3 | 10 | 1112 | |||
GenBank mRNA | AK013540.1 | - | 1222 | |||
AK031242.1 | - | 10 | 1078 | |||
AK031248.1 | - | 10 | 1083 | |||
AK147784.1 | - | 11 | 1118 | |||
AK160881.1 | - | 11 | 1304 | |||
BC003224.1 | - | 8 | 910 | 3482729 | ||
JN086338.1 | - | 9 | 1033 | |||
JN086339.1 | - | 10 | 1271 |