Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 15 | 2622 | ||
NM_134083.4 | NP_598844.3 | 15 | 2617 | |||
GenBank mRNA | AK031242.1 | - | 15 | 2583 | ||
AK147784.1 | - | 16 | 2623 | |||
AK160881.1 | - | 16 | 2809 |