Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134160.1 | NP_598921.1 | 2 | 343 | ||
GenBank mRNA | AF475086.1 | - | 2 | 343 | ||
AK030819.1 | - | 2 | 463 | |||
AK033008.1 | - | 2 | 457 | |||
AK035029.1 | - | 2 | 464 | |||
AK191060.1 | - | 2 | 224 | |||
AY083531.1 | - | 2 | 341 | |||
BC106856.2 | - | 2 | 343 | 40047039 | ||
BC106857.2 | - | 2 | 343 | 40047042 |