Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134191.1 | NP_598952.1 | 1 | 459 | ||
GenBank mRNA | BC119429.1 | - | 1 | 457 | ||
BC119430.1 | - | 1 | 459 | 40040474 |