Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138650.2 | NP_619591.1 | 4, 5 | 763 | ||
GenBank mRNA | AK083258.1 | - | 4, 5 | 756 | ||
AK087972.1 | - | 4, 5 | 721 | |||
AK089261.1 | - | 4, 5 | 561 | |||
AK220294.1 | - | 4, 5 | 705 | |||
BC015278.1 | - | 3, 4 | 336 | 4221290 |