Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138756.4 | NP_620095.1 | 6 | 823 | ||
GenBank mRNA | AK034423.1 | - | 2 | 359 | ||
AK042075.1 | - | 6 | 912 | |||
AK053646.1 | - | 6 | 939 | |||
AK078779.1 | - | 6 | 824 | |||
BC008171.1 | - | 6 | 815 | 3710468 |